FRO182845853 Frontier Scientific Services Inc.

(4-Methoxy-Benzyl)-(2-Morpholin-4-Yl-Ethyl)-Amine

  • MDL number MFCD03724749
  • Molecular Weight : 250.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart