FRO183931648 Frontier Scientific Services Inc.

4-BROMOPHENYL CHLOROFORMATE

  • MDL number MFCD00013256
  • Molecular Weight : 235.46573

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart