FRO183932157 Frontier Scientific Services Inc.

1,5-DI(CHLOROMETHYL)-2,4-DIMETHYLBENZENE

  • MDL number MFCD00013680
  • Molecular Weight : 203.1131

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data FREE
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart