FRO183932934 Frontier Scientific Services Inc.

2-(Piperidin-3-Yl-Methyl)Isoquinolin-1(2H)-One Hydrochloride

  • MDL number MFCD11656753
  • Molecular Weight : 278.7735

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart