FRO183933455 Frontier Scientific Services Inc.

4-Chloro-6,7-Dihydro-2-(Methylthio)-7-Phenyl-(5H)-Pyrrolo[2,3-D]Pyrimidine

  • MDL number MFCD00202984
  • Molecular Weight : 277.7777

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart