FRO183933456 Frontier Scientific Services Inc.

3-Amino-3-Pyridin-3-Ylpropionic Acid Amide

  • MDL number MFCD06216308
  • Molecular Weight : 165.1964

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart