FRO183933522 Frontier Scientific Services Inc.

1-Cyclopentylpiperazin-2-One Hydrochloride

  • Cas Number : 59702-17-9
  • Molecular Weight : 213.6907

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart