FRO183933630 Frontier Scientific Services Inc.

2-(4-EthYl-Benzoyl)Benzoic Acid

  • MDL number MFCD00020288
  • Cas Number : 1151-14-0
  • Molecular Weight : 254.2882

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart