FRO183933632 Frontier Scientific Services Inc.

3-Methyl-Bicyclo(2.2.1)Heptyl-2-Methanol

  • MDL number MFCD00167581
  • Cas Number : 6968-75-8
  • Molecular Weight : 140.2273

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart