FRO183933820 Frontier Scientific Services Inc.

2-Chloroethyl Acetate

  • MDL number MFCD00018963
  • Cas Number : 542-58-5
  • Molecular Weight : 122.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart