FRO183934045 Frontier Scientific Services Inc.

(1-Butyl-1H-Benzoimidazol-2-Yl)-Hydrazine Hydrochloride

  • MDL number MFCD12022653
  • Molecular Weight : 240.738

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart