FRO183934046 Frontier Scientific Services Inc.

[2-(1H-Indol-3-Yl)-Ethyl]-Hydrazine Maleate

  • MDL number MFCD07786368
  • Cas Number : 21216-31-9
  • Molecular Weight : 319.3602

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Maleate
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart