FRO183934133 Frontier Scientific Services Inc.

N-(2-Hydroxypropyl)Ethylenediamine

  • MDL number MFCD00025598
  • Molecular Weight : 118.1801

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart