FRO196829480 Frontier Scientific Services Inc.

3,4-Dichloro-N-EthYl-Benzylamine

  • MDL number MFCD00045168
  • Molecular Weight : 204.1007

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart