FRO196829487 Frontier Scientific Services Inc.

2-[(7-Methyl-2,3-Dihydro-1H-Inden-4-Yl)Oxy]Pyridin-3-Amine

  • MDL number MFCD00098320
  • Cas Number : 175136-11-5
  • Molecular Weight : 240.3076

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart