FRO196829488 Frontier Scientific Services Inc.

3-[(4-PyridYl-Methyl)Amino]Propanenitrile

  • MDL number MFCD00121809
  • Molecular Weight : 161.2081

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart