FRO196829493 Frontier Scientific Services Inc.

3-(Difluoromethoxy)Benzyl Alcohol

  • MDL number MFCD00236227
  • Molecular Weight : 174.1486

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart