FRO196829507 Frontier Scientific Services Inc.

N,N-Diethyl-2-(Piperidin-3-Yl-Oxy)-Acetamide

  • MDL number MFCD08061065
  • Molecular Weight : 214.3102

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart