FRO196829520 Frontier Scientific Services Inc.

2-P-Tolyl-Propionaldehyde

  • MDL number MFCD00072170
  • Molecular Weight : 148.2065

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart