FRO196829551 Frontier Scientific Services Inc.

2-N-Tert-Butyl-6-Chloro-4-N-Cyclopropyl-1,3,5-Triazine-2,4-Diamine

  • MDL number MFCD00155445
  • Cas Number : 32889-48-8
  • Molecular Weight : 241.721

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart