FRO196829560 Frontier Scientific Services Inc.

Ethyl 3-Chloro-6-Phenylpyridazine-4-Carboxylate

  • MDL number MFCD00974814
  • Molecular Weight : 262.692

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart