FRO196829611 Frontier Scientific Services Inc.

Methyl (2S)-2-Amino-3-[4-(Tert-Butoxy)Phenyl]Propanoate Hydrochloride

  • MDL number MFCD00153466
  • Cas Number : 51482-39-4
  • Molecular Weight : 251.3214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart