FRO196829682 Frontier Scientific Services Inc.

2,4-Dimethyl-6-(1-Methylcyclohexyl)Phenol

  • MDL number MFCD00151817
  • Molecular Weight : 218.342

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart