FRO196829686 Frontier Scientific Services Inc.

Phenyl 4-Aminobenzoate

  • MDL number MFCD00189396
  • Molecular Weight : 213.23811

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart