FRO196829718 Frontier Scientific Services Inc.

1-(4-Ethoxyphenyl)Acetone

  • MDL number MFCD00210427
  • Molecular Weight : 178.23303

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart