FRO196829719 Frontier Scientific Services Inc.

1-(3,4,5-Trimethoxyphenyl)Acetone

  • MDL number MFCD00210454
  • Molecular Weight : 224.25893

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart