FRO196829726 Frontier Scientific Services Inc.

(2-Ethyl-6-Methylphenyl)Hydrazine

  • MDL number MFCD02646642
  • Molecular Weight : 150.22533

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Free
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart