FRO196829744 Frontier Scientific Services Inc.

(4-Sec-Butylphenyl)Hydrazine Hydrochloride

  • MDL number MFCD05256416
  • Molecular Weight : 200.70242

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Salt Data Hydrochloride
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart