FRO310396128 Frontier Scientific Services Inc.

(5,6-dimethyl-1H-1,3-benzodiazol-2-yl)methanol

  • MDL number MFCD00022679
  • Molecular Weight : 23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart