FRO310396139 Frontier Scientific Services Inc.

1-(2,4,6-trimethylpyridin-3-yl)ethan-1-ol

  • MDL number MFCD00023508
  • Molecular Weight : 27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart