FRO310396477 Frontier Scientific Services Inc.

N'-hydroxy-4-methanesulfonamidobenzene-1-carboximidamide

  • MDL number MFCD00205140
  • Molecular Weight : 229.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart