FRO333427172 Frontier Scientific Services Inc.

Phthalocyanine

  • MDL number MFCD00005085
  • Cas Number : 574-93-6
  • Molecular Weight : 516.555

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart