FRO333427192 Frontier Scientific Services Inc.

Protoporphyrin IX

  • MDL number MFCD00151109
  • Cas Number : 553-12-8
  • Molecular Weight : 562.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart