FRO333427261 Frontier Scientific Services Inc.

delta-Aminolevulinic acid hydrochloride

  • MDL number MFCD00012869
  • Cas Number : 5451-09-2
  • Molecular Weight : 167.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart