FRO333427299 Frontier Scientific Services Inc.

meso-Tetra(4-sulfonatophenyl)porphine dihydrochloride

  • MDL number MFCD00661071
  • Molecular Weight : 1007.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart