FRO333428446 Frontier Scientific Services Inc.

Dichloro-bis(triphenylphosphine)palladium (II)

  • MDL number MFCD00009593
  • Cas Number : 13965-03-2
  • Molecular Weight : 701.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart