FRO333428990 Frontier Scientific Services Inc.

3-Phenylpropylboronic acid

  • MDL number MFCD01074582
  • Cas Number : 36329-85-8
  • Molecular Weight : 164.009

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart