FRO333429727 Frontier Scientific Services Inc.

Lithium (isoquinolin-1-yl)trihydroxyborate

  • MDL number MFCD18070962
  • Cas Number : 1393823-03-4
  • Molecular Weight : 196.925

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart