FRO386351513 Frontier Scientific Services Inc.

2-(tert-Butoxycarbonylamino)pyridin-4-ylboronic acid

  • MDL number MFCD13188969
  • Cas Number : 903513-59-7
  • Molecular Weight : 238.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart