FRO424040739 Frontier Scientific Services Inc.

5-Ethynyl-3-fluoro-2-methoxypyridine

  • MDL number MFCD18254444
  • Molecular Weight : 151.138

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart