FRO432441458 Frontier Scientific Services Inc.

5-Bromo-2-(chloromethyl)-3-methylpyridine

  • MDL number MFCD18254357
  • Cas Number : 1015060-31-7
  • Molecular Weight : 220.494

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart