FRO447919201 Frontier Scientific Services Inc.

1-(5-Bromopyridin-2-yl)-2,2,2-trifluoroethanone

  • MDL number MFCD07375064
  • Cas Number : 886364-50-7
  • Molecular Weight : 254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart