FRO447919209 Frontier Scientific Services Inc.

2-Methyl-4-(trifluoromethyl)benzyl chloride

  • MDL number MFCD18399883
  • Molecular Weight : 208.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart