FRO447919211 Frontier Scientific Services Inc.

1-(3-Bromofuran-2-yl)-2,2,2-trifluoroethanone

  • MDL number MFCD19613495
  • Molecular Weight : 242.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart