FRO490924745 Frontier Scientific Services Inc.

Tert-butyl bis(4-hydroxybutyl)carbamate

  • MDL number MFCD28053466
  • Cas Number : 1056441-60-1
  • Molecular Weight : 261.358

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart