FRO638474739 Frontier Scientific Services Inc.

1-Boc-3,6-dihydro-2H-pyridine-5-boronic acid pinacol ester

  • MDL number MFCD10697911
  • Cas Number : 885693-20-9
  • Molecular Weight : 295.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart