FROH0399C60E Frontier Scientific Services Inc.

Sodium choleate

  • Cas Number : 361-09-1
  • Molecular Weight : 430.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name sodium 3,7,12-trihydroxycholan-24-oate
Inchi Code 1S/C24H40O5.Na/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26;/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29);/q;+1/p-1
Inchi Key NRHMKIHPTBHXPF-UHFFFAOYSA-M

Description

Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart