FROH0399CA24 Frontier Scientific Services Inc.

2,4-dichloro-1-isothiocyanatobenzene

  • Cas Number : 6590-96-1
  • Molecular Weight : 204.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart