FROH0399DCA4 Frontier Scientific Services Inc.

3-Fluoro-2-methoxypyridine-5-boronic acid pinacol este

  • MDL number MFCD07374975
  • Cas Number : 1310384-35-0
  • Molecular Weight : 253.078

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart