FROH0399E121 Frontier Scientific Services Inc.

(4-(Methoxycarbonyl)-phenyl) zinc pivalate (0.88 mmol/g)

  • Cas Number : 1383708-41-5
  • Molecular Weight : 273.663

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart